1H-Pyrazole-4-boronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-8851-5G
PN-8851-1G
COBBPN-8851-5GEA
0
GBP
COBBPN-8851-5G
COBBPN-8851-1G
1H-Pyrazole-4-boronic acid pinacol ester
1H-Pyrazole-4-boronic acid pinacol ester
Formula:
C₉H₁₅BN₂O₂ |
MDL Number:
MFCD03453063 CAS Number: 269410-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...