2-Aminoimidazole hemisulphate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Aminoimidazole sulphate, 1H-imidazol-2-amine sulphate(2:1)

013113-5G 013113-25G
FLUO013113-5GEA 0 GBP
FLUO013113-5G FLUO013113-25G
2-Aminoimidazole hemisulphate
2-Aminoimidazole hemisulphate
Formula: C₆H₁₂N₆O₄S
MDL Number: MFCD00013162
CAS Number: 1450-93-7
EINECS: 215-918-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR