3-Chloro-6-(1-imidazolyl)pyridazine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-6059-5G
HC-6059-1G
COBBHC-6059-5GEA
0
GBP
COBBHC-6059-5G
COBBHC-6059-1G
3-Chloro-6-(1-imidazolyl)pyridazine
3-Chloro-6-(1-imidazolyl)pyridazine
Formula:
C₇H₅ClN₄ |
MDL Number:
MFCD02323226 CAS Number: 71037-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...