6-Hydroxy-5-(tetrahydropyran-2-yl)-2-naphthaleneboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-5649-1G BB-5649-250M
COBBBB-5649-1GEA 0 GBP
COBBBB-5649-1G COBBBB-5649-250M
6-Hydroxy-5-(tetrahydropyran-2-yl)-2-naphthaleneboronic acid
6-Hydroxy-5-(tetrahydropyran-2-yl)-2-naphthaleneboronic acid
Formula: C₁₅H₁₇BO₄
MDL Number: MFCD11855850
CAS Number: 1150114-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR