6-Methylimidazo[1,2-a]pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1376-5G
HI-1376-25G
HI-1376-1G
COBBHI-1376-5GEA
0
GBP
COBBHI-1376-5G
COBBHI-1376-25G
COBBHI-1376-1G
6-Methylimidazo[1,2-a]pyridine
6-Methylimidazo[1,2-a]pyridine
Formula:
C₈H₈N₂ MW: 132.1625 g/mol |
MDL Number:
MFCD05663806 CAS Number: 874-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...