3-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2130-5G HC-2130-1G
COBBHC-2130-5GEA 0 GBP
COBBHC-2130-5G COBBHC-2130-1G
3-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
3-Bromo-6-methylimidazo[1,2-a]pyridine hydrochloride
Formula: C₈H₈BrClN₂
MDL Number: MFCD11504902
CAS Number: 1072944-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR