6-Bromo-7-methylimidazo[1,2-a]pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HI-1295-5G HI-1295-1G
COBBHI-1295-5GEA 0 GBP
COBBHI-1295-5G COBBHI-1295-1G
6-Bromo-7-methylimidazo[1,2-a]pyridine
6-Bromo-7-methylimidazo[1,2-a]pyridine
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD09258769
CAS Number: 116355-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR