3-Bromo-7-methylimidazo[1,2-a]pyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2143-5G
HC-2143-1G
COBBHC-2143-5GEA
0
GBP
COBBHC-2143-5G
COBBHC-2143-1G
3-Bromo-7-methylimidazo[1,2-a]pyridine
3-Bromo-7-methylimidazo[1,2-a]pyridine
Formula:
C₈H₇BrN₂ MW: 211.0600 g/mol |
MDL Number:
MFCD08056208 CAS Number: 56051-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...