7-Chloro-3-nitroimidazo[1,2-a]pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2070-5G HC-2070-1G
COBBHC-2070-5GEA 0 GBP
COBBHC-2070-5G COBBHC-2070-1G
7-Chloro-3-nitroimidazo[1,2-a]pyridine
7-Chloro-3-nitroimidazo[1,2-a]pyridine
Formula: C₇H₄ClN₃O₂
MDL Number: MFCD11504888
CAS Number: 1072944-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR