4-Chloroimidazo[4,5-c]pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Chloro-1H-imidazo[4,5-c]pyridine
AS5138G1
AS5138G5
ACTIAS5138G1EA
0
GBP
ACTIAS5138G1
ACTIAS5138G5
4-Chloroimidazo[4,5-c]pyridine
4-Chloroimidazo[4,5-c]pyridine
Formula:
C₆H₄ClN₃ |
MDL Number:
MFCD08689582 CAS Number: 2770-01-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...