6-Bromoimidazo[1,2-a]pyridine hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6078-5G HC-6078-25G HC-6078-1G
COBBHC-6078-5GEA 0 GBP
COBBHC-6078-5G COBBHC-6078-25G COBBHC-6078-1G
6-Bromoimidazo[1,2-a]pyridine hydrochloride
6-Bromoimidazo[1,2-a]pyridine hydrochloride
Formula: C₇H₆BrClN₂
MDL Number: MFCD09475874
CAS Number: 957035-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR