6-Chloro-3-nitroimidazo[1,2-b]pyridazine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7804-1G PY-7804-25G PY-7804-5G
COBBPY-7804-1GEA 0 GBP
COBBPY-7804-1G COBBPY-7804-25G COBBPY-7804-5G
6-Chloro-3-nitroimidazo[1,2-b]pyridazine
6-Chloro-3-nitroimidazo[1,2-b]pyridazine
Formula: C₆H₃ClN₄O₂
MDL Number: MFCD08276991
CAS Number: 18087-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR