1-Vinylimidazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

235466-100G 235466-25G
SIAL235466-100GEA 88.6 GBP
SIAL235466-100G SIAL235466-25G
1-Vinylimidazole, Sigma-Aldrich®
1-Vinylimidazole
Formula: C₅H₆N₂
MDL Number: MFCD00005297
CAS Number: 1072-63-5
EINECS: 214-012-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR