3-Pyrid-2-ylbenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Pyridin-2-yl)benzoic acid

AS30228G1 AS30228G5
ACTIAS30228G1EA 0 GBP
ACTIAS30228G1 ACTIAS30228G5
3-Pyrid-2-ylbenzoic acid
3-Pyrid-2-ylbenzoic acid
Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
MDL Number: MFCD05864807
CAS Number: 4467-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR