1-Pyrenebutyric acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
257354-1G
257354-5G
SIAL257354-1GEA
102
GBP
SIAL257354-1G
SIAL257354-5G
1-Pyrenebutyric acid, Sigma-Aldrich®
1-Pyrenebutyric acid
Formula:
C₂₀H₁₆O₂ MW: 288.35 g/mol |
MDL Number:
MFCD00004141 CAS Number: 3443-45-6 EINECS: 222-354-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...