4-(Trifluoromethyl)benzenesulphonyl chloride
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20098G100
AS20098G25
ACTIAS20098G100EA
0
GBP
ACTIAS20098G100
ACTIAS20098G25
4-(Trifluoromethyl)benzenesulphonyl chloride
4-(Trifluoromethyl)benzenesulphonyl chloride
Formula:
C₇H₄ClF₃O₂S Boiling Pt: 49.5…50.5 °C (0.2 mmHg) Melting Pt: 31…33 °C |
MDL Number:
MFCD00042422 CAS Number: 2991-42-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...