4-Cyano-2-fluorobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Fluoro-4-formylbenzonitrile

AS31130G1 AS31130G5
ACTIAS31130G1EA 0 GBP
ACTIAS31130G1 ACTIAS31130G5
4-Cyano-2-fluorobenzaldehyde
4-Cyano-2-fluorobenzaldehyde
Formula: C₈H₄FNO
MW: 149.1200 g/mol
MDL Number: MFCD07368786
CAS Number: 105942-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR