Methyl-4-phenylbenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018823-5G 018823-10G
FLUO018823-5GEA 0 GBP
FLUO018823-5G FLUO018823-10G
Methyl-4-phenylbenzoate
Methyl 4-phenylbenzoate
Formula: C₁₄H₁₂O₂
MDL Number: MFCD00017200
CAS Number: 720-75-2
EINECS: 211-954-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR