Methyl arachidate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl eicosanoate, Methyl Arachidate

M-Clarity™ quality level = MQ200

A3881-1G A3881-25G A3881-5G
SIALA3881-1GEA 95.6 GBP
SIALA3881-1G SIALA3881-25G SIALA3881-5G
Methyl arachidate, Sigma-Aldrich®
Methyl arachidate
Formula: C₂₁H₄₂O₂
MW: 326.56 g/mol
MDL Number: MFCD00009014
CAS Number: 1120-28-1
EINECS: 214-304-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR