2-Amino-6-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20996G1 AS20996G25 AS20996G5
ACTIAS20996G1EA 0 GBP
ACTIAS20996G1 ACTIAS20996G25 ACTIAS20996G5
2-Amino-6-methoxypyridine
2-Amino-6-methoxypyridine
Formula: C₆H₈N₂O
MDL Number: MFCD04972542
CAS Number: 17920-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR