2-Amino-5-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7710-5G PY-7710-1G
COBBPY-7710-5GEA 0 GBP
COBBPY-7710-5G COBBPY-7710-1G
2-Amino-5-methoxypyridine
2-Amino-5-methoxypyridine
Formula: C₆H₈N₂O
MW: 124.1405 g/mol
MDL Number: MFCD07374873
CAS Number: 10167-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR