2-Amino-5-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21843G1 AS21843G5
ACTIAS21843G1EA 0 GBP
ACTIAS21843G1 ACTIAS21843G5
2-Amino-5-methoxypyridine
2-Amino-5-methoxypyridine
Formula: C₆H₈N₂O
MDL Number: MFCD07374873
CAS Number: 10167-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR