2-Amino-4-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20732G1 AS20732G25 AS20732G5
ACTIAS20732G1EA 0 GBP
ACTIAS20732G1 ACTIAS20732G25 ACTIAS20732G5
2-Amino-4-methoxypyridine
2-Amino-4-methoxypyridine
Formula: C₆H₈N₂O
MDL Number: MFCD07437849
CAS Number: 10201-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR