Ethyl dibromofluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003217-25G 003217-100G
FLUO003217-25GEA 0 GBP
FLUO003217-25G FLUO003217-100G
Ethyl dibromofluoroacetate
Ethyl dibromofluoroacetate
Formula: C₄H₅Br₂FO₂
MDL Number: MFCD00042068
CAS Number: 565-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR