6-Nitro-o-cresol
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Methyl-6-nitrophenol
AS21656G1
AS21656G5
ACTIAS21656G1EA
0
GBP
ACTIAS21656G1
ACTIAS21656G5
6-Nitro-o-cresol
6-Nitro-o-cresol
Formula:
C₇H₇NO₃ MW: 153.14 g/mol |
MDL Number:
MFCD00031116 CAS Number: 13073-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...