1-Chloro-2-(trifluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7676-5G OS-7676-25G OS-7676-100G
COBBOS-7676-5GEA 0 GBP
COBBOS-7676-5G COBBOS-7676-25G COBBOS-7676-100G
1-Chloro-2-(trifluoromethoxy)benzene
1-Chloro-2-(trifluoromethoxy)benzene
Formula: C₇H₄ClF₃O
MDL Number: MFCD03840206
CAS Number: 450-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR