3-Amino-2-bromo-6-methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-6-methoxypyridin-3-amine

HC-2077-1G HC-2077-5G
COBBHC-2077-1GEA 0 GBP
COBBHC-2077-1G COBBHC-2077-5G
3-Amino-2-bromo-6-methoxypyridine
3-Amino-2-bromo-6-methoxypyridine
Formula: C₆H₇BrN₂O
MW: 203.0400 g/mol
MDL Number: MFCD05662394
CAS Number: 135795-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR