2-(Cyanomethoxy)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-9003-1G BB-9003-250M
COBBBB-9003-1GEA 0 GBP
COBBBB-9003-1G COBBBB-9003-250M
2-(Cyanomethoxy)phenylboronic acid
2-(Cyanomethoxy)phenylboronic acid
Formula: C₈H₈BNO₃
MDL Number: MFCD09908171
CAS Number: 947533-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR