5-(Trifluoromethoxy)salicylaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde

005558-1G 005558-5G
FLUO005558-1GEA 0 GBP
FLUO005558-1G FLUO005558-5G
5-(Trifluoromethoxy)salicylaldehyde
5-(Trifluoromethoxy)salicylaldehyde
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
MDL Number: MFCD00075249
CAS Number: 93249-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR