Ethyl iodofluoroacetate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009177-5G
009177-25G
FLUO009177-5GEA
0
GBP
FLUO009177-5G
FLUO009177-25G
Ethyl iodofluoroacetate
Ethyl iodofluoroacetate
Formula:
C₄H₆FIO₂ |
MDL Number:
MFCD01320733 CAS Number: 401-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...