Ethyl iododifluoroacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2,2-difluoro-2-iodoacetate, Ethyl difluoro(iodo)acetate

010300-5G 010300-25G
FLUO010300-5GEA 0 GBP
FLUO010300-5G FLUO010300-25G
Ethyl iododifluoroacetate
Ethyl iododifluoroacetate
Formula: C₄H₅F₂IO₂
MDL Number: MFCD00798135
CAS Number: 7648-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR