1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: O-(4-(Trifluoromethyl)benzyl)hydroxylamine hydrochloride

010012-1G 010012-5G
FLUO010012-1GEA 0 GBP
FLUO010012-1G FLUO010012-5G
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride
1-[(Ammoniooxy)methyl]-4-(trifluoromethyl)benzene chloride
Formula: C₈H₉ClF₃NO
MDL Number: MFCD00231881
CAS Number: 321574-29-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR