1-(Methylsulphonyl)piperazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20639G25 AS20639G5
ACTIAS20639G25EA 0 GBP
ACTIAS20639G25 ACTIAS20639G5
1-(Methylsulphonyl)piperazine
1-(Methylsulphonyl)piperazine
Formula: C₅H₁₂N₂O₂S
MDL Number: MFCD02153225
CAS Number: 55276-43-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR