(±)-2-Piperazinecarboxylic acid dihydrochloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Piperazine-2-carboxylic acid dihydrochloride
019229-1G
019229-25G
019229-5G
FLUO019229-1GEA
0
GBP
FLUO019229-1G
FLUO019229-25G
FLUO019229-5G
(±)-2-Piperazinecarboxylic acid dihydrochloride
(±)-2-Piperazinecarboxylic acid dihydrochloride
Formula:
C₅H₁₂Cl₂N₂O₂ MW: 203.07 g/mol |
MDL Number:
MFCD00044803 CAS Number: 3022-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...