1-(2-Cyclohexylethyl)piperazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008915-1G
008915-5G
FLUO008915-1GEA
0
GBP
FLUO008915-1G
FLUO008915-5G
1-(2-Cyclohexylethyl)piperazine
1-(2-Cyclohexylethyl)piperazine
Formula:
C₁₂H₂₄N₂ |
MDL Number:
MFCD01320901 CAS Number: 132800-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...