Arachidonic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (5Z,8Z,11Z,14Z)-Icosa-5,8,11,14-tetraenoic acid

A3611-100MG A3611-1G
SIALA3611-100MGEA 243 GBP
SIALA3611-100MG SIALA3611-1G
Arachidonic acid, Sigma-Aldrich®
Arachidonic acid
Formula: C₂₀H₃₂O₂
MW: 304.5 g/mol
MDL Number: MFCD00004417
CAS Number: 506-32-1
EINECS: 208-033-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR