Estradiol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

E1024-100G E1024-1G E1024-25G
SIALE1024-100GEA 4780 GBP
SIALE1024-100G SIALE1024-1G SIALE1024-25G
Estradiol, Sigma-Aldrich®
Estradiol
Formula: C₁₈H₂₄O₂
MDL Number: MFCD00003693
CAS Number: 50-28-2
EINECS: 200-023-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR