(R)-(+)-1-Boc-3-aminopyrrolidine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (R)-1-Boc-3-aminopyrrolidine, (3R)-(+)-1-Boc-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-Boc-pyrrolidine, (R)-tert-Butyl 3-aminopyrrolidine-1-carboxylate, (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, (3R)-(+)-3-Amino-1-(tert-butoxycarbonyl)pyrrolidine

011315-1G 011315-5G
FLUO011315-1GEA 0 GBP
FLUO011315-1G FLUO011315-5G
(R)-(+)-1-Boc-3-aminopyrrolidine
(R)-(+)-1-Boc-3-aminopyrrolidine
Formula: C₉H₁₈N₂O₂
MDL Number: MFCD03419272
CAS Number: 147081-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR