Thiazole-2-carbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Thiazolecarboxaldehyde, Thiazole-2-carboxaldehyde

AS50348G1 AS50348G100 AS50348G5
ACTIAS50348G1EA 0 GBP
ACTIAS50348G1 ACTIAS50348G100 ACTIAS50348G5
Thiazole-2-carbaldehyde
Thiazole-2-carbaldehyde
Formula: C₄H₃NOS
MW: 113.13 g/mol
MDL Number: MFCD00142924
CAS Number: 10200-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR