3'-Methylpropiophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21144G25 AS21144G5
ACTIAS21144G25EA 185 GBP
ACTIAS21144G25 ACTIAS21144G5
3'-Methylpropiophenone
3'-Methylpropiophenone
Formula: C₁₀H₁₂O
MW: 148.2017 g/mol
MDL Number: MFCD00027014
CAS Number: 51772-30-6
EINECS: 257-405-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR