7-Amino-1-tetralone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21506G1 AS21506G5
ACTIAS21506G1EA 0 GBP
ACTIAS21506G1 ACTIAS21506G5
7-Amino-1-tetralone
7-Amino-1-tetralone
Formula: C₁₀H₁₁NO
MDL Number: MFCD07369817
CAS Number: 22009-40-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR