3,5-Dimethyl-4-iodo-1H-pyrazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethyl-4-iodopyrazole

AS22163G100 AS22163G25
ACTIAS22163G100EA 122 GBP
ACTIAS22163G100 ACTIAS22163G25
3,5-Dimethyl-4-iodo-1H-pyrazole
3,5-Dimethyl-4-iodo-1H-pyrazole
Formula: C₅H₇IN₂
MW: 222.03 g/mol
MDL Number: MFCD00040247
CAS Number: 2033-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR