Ferrocenecarboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS24273G100 AS24273G25
ACTIAS24273G100EA 0 GBP
ACTIAS24273G100 ACTIAS24273G25
Ferrocenecarboxylic acid
Ferrocenecarboxylic acid
Formula: C₁₁H₁₀FeO₂
MDL Number: MFCD00001430
CAS Number: 1271-42-7
EINECS: 215-040-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR