1,1'-Bis(diphenylphosphino)ferrocene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: DPPF, 1,1-Bis(diphenylphosphino)ferrocene

AS20195G100 AS20195G25 AS20195G5
ACTIAS20195G100EA 0 GBP
ACTIAS20195G100 ACTIAS20195G25 ACTIAS20195G5
1,1'-Bis(diphenylphosphino)ferrocene
1,1'-Bis(diphenylphosphino)ferrocene
Formula: C₃₄H₂₈FeP₂
MW: 554.39 g/mol
MDL Number: MFCD00001422
CAS Number: 12150-46-8
EINECS: 430-420-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR