2-Bromo-5-thiazolecarboxylic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

016153-1G 016153-5G
FLUO016153-1GEA 0 GBP
FLUO016153-1G FLUO016153-5G
2-Bromo-5-thiazolecarboxylic acid
2-Bromo-5-thiazolecarboxylic acid
Formula: C₄H₂BrNO₂S
MDL Number: MFCD04115730
CAS Number: 54045-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR