2,4'-Difluorobenzophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-Fluorophenyl)(4-fluorophenyl)methanone

AS23121G100 AS23121G25
ACTIAS23121G100EA 75 GBP
ACTIAS23121G100 ACTIAS23121G25
2,4'-Difluorobenzophenone
2,4'-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 22…24 °C
Density: 1,24 g/cm³ (20 °C)
MDL Number: MFCD00000319
CAS Number: 342-25-6
EINECS: 206-441-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR