7-Azaindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS7179G100 AS7179G25 AS7179G5
ACTIAS7179G100EA 0 GBP
ACTIAS7179G100 ACTIAS7179G25 ACTIAS7179G5
7-Azaindole
7-Azaindole
Formula: C₇H₆N₂
Boiling Pt: 384 °C (1013 hPa)
Melting Pt: 103…106 °C
Density: 1,301 g/cm³ (20 °C)
MDL Number: MFCD00005606
CAS Number: 271-63-6
EINECS: 205-981-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR