6-Azaindole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2957G1 AS2957G25 AS2957G5
ACTIAS2957G1EA 0 GBP
ACTIAS2957G1 ACTIAS2957G25 ACTIAS2957G5
6-Azaindole
6-Azaindole
Formula: C₇H₆N₂
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR