5-Azaindole
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine
AS7202G1
AS7202G25
AS7202G5
ACTIAS7202G1EA
0
GBP
ACTIAS7202G1
ACTIAS7202G25
ACTIAS7202G5
5-Azaindole
5-Azaindole
Formula:
C₇H₆N₂ MW: 118.14 g/mol |
MDL Number:
MFCD00955936 CAS Number: 271-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...