1-(4-Bromophenyl)-2-thiourea

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-(4-Bromophenyl)thiourea, N-(4-Bromophenyl)thiourea, 4-Bromophenylthiourea

OT-0903-25G OT-0903-5G
COBBOT-0903-25GEA 0 GBP
COBBOT-0903-25G COBBOT-0903-5G
1-(4-Bromophenyl)-2-thiourea
1-(4-Bromophenyl)-2-thiourea
Formula: (C₆H₄Br)NHC(S)NH₂
MW: 231.12 g/mol
Boiling Pt: 319 °C (1013 hPa)
Melting Pt: 186 °C
MDL Number: MFCD00041176
CAS Number: 2646-30-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR